Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-00092 NBP1-00092SS
NOVUNBP1-00092EA 599 GBP
NOVUNBP1-00092 NOVUNBP1-00092SS
Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Macrophage Scavenger Receptor I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR