Anti-HIST3H3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB500-171-A NB500-171-B
NOVUNB500-171-AEA 642 GBP
NOVUNB500-171-A NOVUNB500-171-B
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Histone H3 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Chicken, Drosophila, Plant, Xenopus.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:300

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR