Anti-MCOLN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-82375 NB110-82375SS
NOVUNB110-82375EA 591 GBP
NOVUNB110-82375 NOVUNB110-82375SS
Anti-MCOLN1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Mucolipin 1 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: MCLN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR