Anti-HAP1 Mouse monoclonal antibody unconjugated [clone: 1B6]

Supplier: Novus Biologicals
NB110-74569 NB110-74569SS
NOVUNB110-74569EA 551 GBP
NOVUNB110-74569 NOVUNB110-74569SS
Anti-HAP1 Mouse monoclonal antibody unconjugated [clone: 1B6]
Antibodies
Mouse Monoclonal HAP1 Antibody (1B6). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 0.5-1.0 ?g/ml

Type: Primary
Antigen: HAP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR