Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]

Supplier: Novus Biologicals
NB110-60011 NB110-60011SS
NOVUNB110-60011EA 603 GBP
NOVUNB110-60011 NOVUNB110-60011SS
Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]
Antibodies
The WT1 Antibody (6F-H2) from Novus Biologicals is a mouse monoclonal antibody to WT1. This antibody reacts with human, mouse. The WT1 Antibody (6F-H2) has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR