Anti-TAZ Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-58359 NB110-58359SS
NOVUNB110-58359EA 573 GBP
NOVUNB110-58359 NOVUNB110-58359SS
Anti-TAZ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TAZ (tafazzin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR