Anti-Tubby Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1922 NB100-1922SS
NOVUNB100-1922EA 539 GBP
NOVUNB100-1922 NOVUNB100-1922SS
Anti-Tubby Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Tubby
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: C. elegans
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR