Anti-IgG Donkey Polyclonal Antibody (CF™ 750)

Supplier: Biotium
20298 20298-1
20298.EA 346 GBP
20298. 20298-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 750)
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 750 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR