Anti-IgG2a Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium
20261 20261-1
20261.EA 370 GBP
20261. 20261-1
Anti-IgG2a Goat Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR