Anti-IgG1 Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20249-1EA 155 GBP
20249-1 20249.
Anti-IgG1 Goat Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR