Anti-IgG Rabbit Polyclonal Antibody (CF™ 350)

Supplier: Biotium
20149 20149-1
20149.EA 346 GBP
20149. 20149-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR