Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium
20147 20147-1
20147.EA 346 GBP
20147. 20147-1
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR