Anti-Lamin B1 Rabbit Monoclonal Antibody [clone: EPR8985(B)]

Supplier: Abcam
AB133741-100 AB133741-20UL AB133741-1ML
ABCAAB133741-100EA 409 GBP
ABCAAB133741-100 ABCAAB133741-20UL ABCAAB133741-1ML
Anti-Lamin B1 Rabbit Monoclonal Antibody [clone: EPR8985(B)]
Antibodies
Rabbit monoclonal [EPR8985(B)] to Lamin B1 - Nuclear Envelope Marker.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Lamin B1
Clonality: Monoclonal
Clone: EPR8985(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR