Anti-Integrin alpha 6 Rabbit Monoclonal Antibody [clone: EPR5578]

Supplier: Abcam
AB124924-100 AB124924-1ML
ABCAAB124924-100EA 613 GBP
ABCAAB124924-100 ABCAAB124924-1ML
Anti-Integrin alpha 6 Rabbit Monoclonal Antibody [clone: EPR5578]
Antibodies
Rabbit monoclonal [EPR5578] to Integrin alpha 6.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Integrin alpha 6
Clonality: Monoclonal
Clone: EPR5578
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR