Anti-Cytokeratin 8 Rabbit Monoclonal Antibody [clone: EP1630]

Supplier: Abcam
AB109452-100 AB109452-1ML
ABCAAB109452-100EA 613 GBP
ABCAAB109452-100 ABCAAB109452-1ML
Anti-Cytokeratin 8 Rabbit Monoclonal Antibody [clone: EP1630]
Antibodies
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: EP1630
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR