Anti-alpha Internexin Rabbit Monoclonal Antibody [clone: EPR1529]

Supplier: Abcam
AB108302-100 AB108302-1ML
ABCAAB108302-100EA 607 GBP
ABCAAB108302-100 ABCAAB108302-1ML
Anti-alpha Internexin Rabbit Monoclonal Antibody [clone: EPR1529]
Antibodies
Rabbit monoclonal [EPR1529] to alpha Internexin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha Internexin
Clonality: Monoclonal
Clone: EPR1529
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR