Anti-Polyethylene glycol Rat Monoclonal Antibody [clone: 26A04]

Supplier: Abcam
AB94764-500 AB94764-100
ABCAAB94764-500EA 1370 GBP
ABCAAB94764-500 ABCAAB94764-100
Anti-Polyethylene glycol Rat Monoclonal Antibody [clone: 26A04]
Antibodies
Rat monoclonal [26A04] to Polyethylene glycol - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Polyethylene glycol
Clonality: Monoclonal
Clone: 26A04
Conjugation:
Epitope:
Host: Rat
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR