Anti-CHEK1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB99396-100 ORB99396-20
BIRBORB99396-100EA 0 GBP
BIRBORB99396-100 BIRBORB99396-20
Anti-CHEK1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR