Anti-RUNX1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB99295-100 ORB99295-20
BIRBORB99295-100EA 0 GBP
BIRBORB99295-100 BIRBORB99295-20
Anti-RUNX1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: RUNX1 (runt-related transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR