Anti-PDPK1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB136061-100 ORB136061-200 ORB136061-50
BIRBORB136061-100EA 0 GBP
BIRBORB136061-100 BIRBORB136061-200 BIRBORB136061-50
Anti-PDPK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDPK1 (3-phosphoinositide dependent protein kinase-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR