Anti-SMAD1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB129744-100 ORB129744-200
BIRBORB129744-100EA 0 GBP
BIRBORB129744-100 BIRBORB129744-200
Anti-SMAD1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMAD1 (SMAD family member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR