Anti-RB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB129734-100 ORB129734-200
BIRBORB129734-100EA 0 GBP
BIRBORB129734-100 BIRBORB129734-200
Anti-RB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR