Anti-WASF1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128912-100 ORB128912-200
BIRBORB128912-100EA 0 GBP
BIRBORB128912-100 BIRBORB128912-200
Anti-WASF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WASF1 (WAS protein family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR