Anti-FGR Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128065-100 ORB128065-200
BIRBORB128065-100EA 0 GBP
BIRBORB128065-100 BIRBORB128065-200
Anti-FGR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGR (feline Gardner-Rasheed sarcoma viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR