Anti-EPHA2/3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128024-100 ORB128024-200
BIRBORB128024-100EA 0 GBP
BIRBORB128024-100 BIRBORB128024-200
Anti-EPHA2/3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EPHA2/3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR