Anti-BSG Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127767-100 ORB127767-200
BIRBORB127767-100EA 0 GBP
BIRBORB127767-100 BIRBORB127767-200
Anti-BSG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR