Anti-RBM39 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127734-100 ORB127734-200
BIRBORB127734-100EA 0 GBP
BIRBORB127734-100 BIRBORB127734-200
Anti-RBM39 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RBM39 (RNA binding motif protein 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR