Anti-ICK Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127410-100 ORB127410-200
BIRBORB127410-100EA 0 GBP
BIRBORB127410-100 BIRBORB127410-200
Anti-ICK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ICK (intestinal cell (MAK-like) kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR