Anti-MKL1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127064-100 ORB127064-200
BIRBORB127064-100EA 0 GBP
BIRBORB127064-100 BIRBORB127064-200
Anti-MKL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MKL1 (megakaryoblastic leukemia (translocation) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR