Anti-MRPL10 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125976-100 ORB125976-200
BIRBORB125976-100EA 0 GBP
BIRBORB125976-100 BIRBORB125976-200
Anti-MRPL10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MRPL10 (mitochondrial ribosomal protein L10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR