Anti-KIR2DL5B Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125909-100 ORB125909-200
BIRBORB125909-100EA 0 GBP
BIRBORB125909-100 BIRBORB125909-200
Anti-KIR2DL5B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KIR2DL5B (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR