Anti-EID1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125702-100 ORB125702-200
BIRBORB125702-100EA 0 GBP
BIRBORB125702-100 BIRBORB125702-200
Anti-EID1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EID1 (EP300 interacting inhibitor of differentiation 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR