Anti-MAPK9 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB106162-100 ORB106162-20
BIRBORB106162-100EA 0 GBP
BIRBORB106162-100 BIRBORB106162-20
Anti-MAPK9 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR