Anti-CBLB Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101851-100 ORB101851-200
BIRBORB101851-100EA 0 GBP
BIRBORB101851-100 BIRBORB101851-200
Anti-CBLB Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: CBLB (Cbl proto-oncogene B, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR