Anti-CPB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101281-100 ORB101281-200
BIRBORB101281-100EA 0 GBP
BIRBORB101281-100 BIRBORB101281-200
Anti-CPB1 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: CPB1 (carboxypeptidase B1 (tissue))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR