Anti-RET Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100148-100 ORB100148-200
BIRBORB100148-100EA 0 GBP
BIRBORB100148-100 BIRBORB100148-200
Anti-RET Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: RET (ret proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR