Anti-TP53 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB22899-0.2 ORB22899-100
BIRBORB22899-0.2EA 0 GBP
BIRBORB22899-0.2 BIRBORB22899-100
Anti-TP53 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR