Anti-NKp44 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB22890-100 ORB22890-20 ORB22890-5
BIRBORB22890-100EA 0 GBP
BIRBORB22890-100 BIRBORB22890-20 BIRBORB22890-5
Anti-NKp44 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: NKp44
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR