Anti-MHC Class Mouse Monoclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB22857-500 ORB22857-1
BIRBORB22857-500EA 0 GBP
BIRBORB22857-500 BIRBORB22857-1
Anti-MHC Class Mouse Monoclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: MHC Class
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR