Anti-Influenza H5N1 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB22722-10 ORB22722-50
BIRBORB22722-10EA 0 GBP
BIRBORB22722-10 BIRBORB22722-50
Anti-Influenza H5N1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Influenza H5N1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR