Anti-FCGR1A Mouse monoclonal antibody

Supplier: Biorbyt
ORB12115-100 ORB12115-500
BIRBORB12115-100EA 0 GBP
BIRBORB12115-100 BIRBORB12115-500
Anti-FCGR1A Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: FCGR1A (Fc fragment of IgG, high affinity Ia, receptor (CD64))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR