Anti-CD48 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB12081-100 ORB12081-50
BIRBORB12081-100EA 0 GBP
BIRBORB12081-100 BIRBORB12081-50
Anti-CD48 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: CD48 (CD48 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR