Anti-IL18 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB10899-100 ORB10899-20 ORB10899-200 ORB10899-5
BIRBORB10899-100EA 0 GBP
BIRBORB10899-100 BIRBORB10899-20 BIRBORB10899-200 BIRBORB10899-5
Anti-IL18 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IL18 (interleukin 18 (interferon-gamma-inducing factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR