Anti-Myosin light chain Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB6428-100 ORB6428-200
BIRBORB6428-100EA 0 GBP
BIRBORB6428-100 BIRBORB6428-200
Anti-Myosin light chain Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Myosin light chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR