Anti-C17orf59 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2804-100 ORB2804-200
BIRBORB2804-100EA 0 GBP
BIRBORB2804-100 BIRBORB2804-200
Anti-C17orf59 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: C17orf59 (chromosome 17 open reading frame 59)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR