Anti-SAMD9 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2643-100 ORB2643-200
BIRBORB2643-100EA 0 GBP
BIRBORB2643-100 BIRBORB2643-200
Anti-SAMD9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SAMD9 (sterile alpha motif domain containing 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR