Anti-AFAP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2045-100 ORB2045-20 ORB2045-200 ORB2045-5
BIRBORB2045-100EA 0 GBP
BIRBORB2045-100 BIRBORB2045-20 BIRBORB2045-200 BIRBORB2045-5
Anti-AFAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: AFAP1 (actin filament associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR