Anti-MIS18BP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1508-100 ORB1508-200
BIRBORB1508-100EA 0 GBP
BIRBORB1508-100 BIRBORB1508-200
Anti-MIS18BP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: MIS18BP1 (MIS18 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR