Anti-Clenbuterol Hydrochloride Rabbit polyclonal antibody

Supplier: Biorbyt
ORB1394-100 ORB1394-200
BIRBORB1394-100EA 0 GBP
BIRBORB1394-100 BIRBORB1394-200
Anti-Clenbuterol Hydrochloride Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: Clenbuterol Hydrochloride
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR