Anti-ABCA7 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1068-100 ORB1068-200
BIRBORB1068-100EA 0 GBP
BIRBORB1068-100 BIRBORB1068-200
Anti-ABCA7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR